Dimethyl sulphoxide-[D₆] (99.9% D) with TMS (0.1 vol.%), MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®
Catalog # 1.03587.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Deuteration degree:≥ 99.9 %
- Water (K. Fischer, H₂O + D₂O):≤ 0.030 %
- Tetramethylsilane (NMR-spectrum):0.001
- Water (NMR, H₂O):≤ 0.020 %
- Performance test (NMR-spectrum):passes test
Specification Test Results
About this item
:
:84.09 g/mol
Boiling Pt:189 °C (5 mmHg)
Melting Pt:20.2 °C
Density:1.19 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:82 °C (closed cup)
Storage Temperature:Ambient
EINECS:218-617-0
CAS Number:2206-27-1
Formula:D₃CS(O)CD₃
MDL Number:MFCD00002090