Dimethyl sulphoxide-[D₆] (99.95% D) with TMS (0.03 vol.%) for NMR spectroscopy, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:2206-27-1
:
Formula:D₃CS(O)CD₃
MDL Number:MFCD00002090
:84.09 g/mol
Boiling Pt:189 °C (5 mmHg)
Melting Pt:20.2 °C
Density:1.19 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:82 °C (closed cup)
Storage Temperature:Ambient
EINECS:218-617-0
Specification Test Results
- Deuteration degree:≥ 99.95 %
- Water - K. Fischer, H₂O + D₂O:≤ 0.02 %
- Performance test (NMR-spectrum):passes test
- Water - NMR, H₂O:≤ 0.01 %
- Tetramethylsilane (NMR-spectrum):≤ 0.03 %
Specification Test Results
Frequently Bought Together