Order Entry
Czech Republic
ContactUsLinkComponent
Toluene-[D₈] 99.5 atom % D + 0.03% (v/v) TMS for NMR spectroscopy
Toluene-[D₈] 99.5 atom % D + 0.03% (v/v) TMS for NMR spectroscopy
Certificates
undefined
Toluene-[D₈] 99.5 atom % D + 0.03% (v/v) TMS for NMR spectroscopy
Certificates

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.03 %
  • D-Enrichment:
    ≥99.4 %

Specification Test Results

Related Information

Frequently Bought Together