

Chloroform-[D₁] for NMR spectroscopy, Sigma-Aldrich®
Catalog # SIAL612200-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:120.38 g/mol
Boiling Pt:60.8 °C (1013 hPa)
Melting Pt:–64.1 °C
Density:1.5 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:212-742-4
UN:1888
ADR:6.1,III
:M-Clarity™ quality level = MQ100
:GHS08
:GHS07
CAS Number:865-49-6
Formula:CDCl₃
MDL Number:MFCD00000827