Order Entry
Czech Republic
ContactUsLinkComponent
Petroleum spirit, 80…110 °C, TECHNICAL
Flammable
Health
Irritant
Environment
Petroleum spirit, 80…110 °C, TECHNICAL
Supplier:  VWR Chemicals
Certificates
undefined
Flammable
Health
Irritant
Environment
Petroleum spirit, 80…110 °C, TECHNICAL
Supplier:  VWR Chemicals
Certificates
Limited Time Sale
Select items below are currently on sale

Specification Test Results

  • Density (20/4):
    0.680 - 0.730
  • Distillation (min. 90 %):
    80 - 110 °C

Specification Test Results

Frequently Bought Together