Order Entry
Czech Republic
ContactUsLinkComponent
Petroleum spirit, 60…100 °C, BAKER, Laboratory Reagent, J.T.Baker®
Flammable
Health
Irritant
Environment
Petroleum spirit, 60…100 °C, BAKER, Laboratory Reagent, J.T.Baker®
Catalog # 8241.9025
Supplier:  Avantor
CAS Number:  
Flammable
Health
Irritant
Environment
Petroleum spirit, 60…100 °C, BAKER, Laboratory Reagent, J.T.Baker®
Catalog # 8241.9025
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    25 L
  • Seq:
    5327

Specifications

Specification Test Results

  • Boiling Range:
    60 - 100 °C
  • Residue after Evaporation:
    max. 0.002 %

Specification Test Results

About this item

 :
Boiling Pt:60…100 °C (1013 hPa)
Density:0.67 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:265-151-9
UN:1268
ADR:3,II
 :GHS02
 :GHS08
 :GHS07
 :GHS09
CAS Number:64742-49-0
 :8008-20-6
MDL Number:MFCD00081849