



Petroleum spirit, 40…80 °C for denaturation, Supelco®
Catalog # 1.01769.1000
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
Boiling Pt:40…70 °C (1013 hPa)
Density:0.666 g/cm³ (15 °C)
UN:1268
ADR:3,II
:M-Clarity™ quality level = MQ200
EINECS:265-151-9
:GHS02
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
CAS Number:64742-49-0
:8008-20-6
MDL Number:MFCD00081849