Order Entry
Czech Republic
ContactUsLinkComponent
Petroleum spirit, 40…60 °C, EMPROVE® ESSENTIAL DAB, SAFC®
Flammable
Health
Irritant
Environment
Petroleum spirit, 40…60 °C, EMPROVE® ESSENTIAL DAB, SAFC®
Catalog # 1.00909.5000
CAS Number:  
Flammable
Health
Irritant
Environment
Petroleum spirit, 40…60 °C, EMPROVE® ESSENTIAL DAB, SAFC®
Catalog # 1.00909.5000
CAS Number:  

Specifications

  • Pk:
    5 L
  • Seq:
    5190

Specifications

About this item

 :
Boiling Pt:40…60 °C (1013 hPa)
Density:0.65 g/cm³ (20 °C)
Flash Pt:–40 °C
Storage Temperature:Ambient
EINECS:265-151-9
UN:1268
ADR:3,I
 :GHS02
 :GHS08
 :GHS07
 :GHS09
CAS Number:64742-49-0
 :101316-46-5
MDL Number:MFCD00081849