Order Entry
Czech Republic
ContactUsLinkComponent
Petroleum spirit, 40…60 °C, BAKER, Laboratory Reagent, J.T.Baker®
Flammable
Health
Irritant
Environment
Petroleum spirit, 40…60 °C, BAKER, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
Certificates
Flammable
Health
Irritant
Environment
Petroleum spirit, 40…60 °C, BAKER, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Appearance:
    passes test
  • Appearance at 0°C:
    passes test
  • Distillation Range:
    40 - 65 °C
  • Residue after Evaporation:
    max. 0.002 %

Specification Test Results

Related Information

Frequently Bought Together