ContactUsLinkComponent
Petroleum spirit, 40…65 °C, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
 :  Avantor
Petroleum spirit, 40…65 °C, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
 :  Avantor
  • Boiling Range:
    40 - 65 °C
  • Residue after Evaporation:
    max. 0.001 %
  • Water (H2O):
    max. 0.02 %
  • Aluminium (Al):
    max. 0.5
  • Boron (B):
    max. 0.02
  • Barium (Ba):
    max. 0.1
  • Calcium (Ca):
    max. 0.5
  • Cadmium (Cd):
    max. 0.05
  • Cobalt (Co):
    max. 0.02
  • Chromium (Cr):
    max. 0.02
  • Copper (Cu):
    max. 0.02
  • Iron (Fe):
    max. 0.1
  • Magnesium (Mg):
    max. 0.1
  • Manganese (Mn):
    max. 0.02
  • Nickel (Ni):
    max. 0.02
  • Lead (Pb):
    max. 0.1
  • Tin (Sn):
    max. 0.1
  • Zinc (Zn):
    max. 0.1
Frequently Bought Together