Order Entry
Czech Republic
ContactUsLinkComponent
Petroleum spirit, 40…60 °C for residue analysis, suitable for PAH analysis
Health
Environment
Flammable
Irritant
Petroleum spirit, 40…60 °C for residue analysis, suitable for PAH analysis
Certificates
undefined
Health
Environment
Flammable
Irritant
Petroleum spirit, 40…60 °C for residue analysis, suitable for PAH analysis

Specification Test Results

  • Appearance:
    Clear liquid
  • Water:
    ≤0.01 % (Coulometric)
  • Boiling point:
    40 to 60 °C
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.0003 %

Specification Test Results

Related Information

Frequently Bought Together