



Petroleum spirit, 40…60 °C ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:64742-49-0
:64742-82-1
MDL Number:MFCD00081849
Boiling Pt:40…60 °C (1013 hPa)
Density:0.65 g/cm³
Flash Pt:–45 °C
Storage Temperature:Ambient
EINECS:265-151-9
UN:1268
ADR:3,II
:GHS08
:GHS09
:GHS02
:GHS07
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.01 % (Coulometric)
- Boiling point:Not less than 90% distils within each range
- Color scale:≤10 APHA
- Residue after evaporation:≤0.001 %
- Acidity/alkalinity:≤0.0001 meq/g corresp. 0.0006%
- Substance darkened by H2SO4:≤10 APHA
- Copper (Cu):≤0.1 ppm
- Iron (Fe):≤0.1 ppm
- Lead (Pb):≤0.1 ppm
- Zinc (Zn):≤0.1 ppm
Specification Test Results
Frequently Bought Together