ContactUsLinkComponent
Molecular sieve 13X (1.0 nm, 10 Å), beads 8-12 mesh, Fluka™
Molecular sieve 13X (1.0 nm, 10 Å), beads 8-12 mesh, Fluka™
Certificates
Molecular sieve 13X (1.0 nm, 10 Å), beads 8-12 mesh, Fluka™
Specification Test Results
  • Form:
    beads
  • Grain size 8 - 12 mesh:
    Min. 95 %