Order Entry
Czech Republic
ContactUsLinkComponent
 
Paraffin, Solidification point: 56…58 °C, in pastille form for histology, Sigma-Aldrich®
Catalog # 1.07164.2504
Supplier:  Merck
CAS Number:  
Paraffin, Solidification point: 56…58 °C, in pastille form for histology, Sigma-Aldrich®
Catalog # 1.07164.2504
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bag
  • Pk:
    4
  • Seq:
    4082
  • Pk Info:
    4 x 2,5 kg (Preparation Capacity 1200 paraffin blocks)

Specifications

Specification Test Results

  • Effect of heating (at 60 °C/100 g):
    passes test
  • Solubility in xylene (at 20 °C/0.1 g in 5 ml):
    passes test
  • Cutting results (4 μm slices):
    passes test
  • Solubility of slices in xylene:
    ≤ 50 s
  • Solidification point (rotating thermometer method):
    56 - 58°C

Specification Test Results

About this item

 :
Density:0.9 g/cm³ (20 °C)
Flash Pt:>100 °C
Storage Temperature:Ambient
EINECS:232-315-6
 :1
CAS Number:64742-51-4
 :8002-74-2