Order Entry
Czech Republic
ContactUsLinkComponent
Molecular sieve 13X (1.0 nm, 10 Å) 8-12 mesh
Molecular sieve 13X (1.0 nm, 10 Å) 8-12 mesh
Certificates
undefined
Molecular sieve 13X (1.0 nm, 10 Å) 8-12 mesh
Certificates

Specification Test Results

  • Appearance:
    Beige to brown Beads
  • Particle size:
    8 to 12 mesh
  • Bulk density:
    ≥600 g/l
  • Residual water:
    ≤1.5 % (550°C, 2 h)
  • CO2 adsorption capacity:
    ≥11.5 %

Specification Test Results

Related Information

Frequently Bought Together