Order Entry
Czech Republic
ContactUsLinkComponent
Paraffin, Solidification point: 52-54°C, pellets, TECHNICAL
Paraffin, Solidification point: 52-54°C, pellets, TECHNICAL
Catalog # 26155.294
Supplier:  VWR Chemicals
CAS Number:  
undefined
Paraffin, Solidification point: 52-54°C, pellets, TECHNICAL
Catalog # 26155.294
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Congealing temperature:
    52 - 54 °C

Specification Test Results

About this item

 :
Melting Pt:52…54 °C
Density:0.9 g/cm³ (20 °C)
EINECS:232-315-6
CAS Number:64742-51-4