2,6-Dimethylcyclohexanone, Sigma-Aldrich®
Catalog # SIAL102261-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:126.2 g/mol
Boiling Pt:174 °C (1013 hPa)
Density:0.92 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:3,III
:M-Clarity™ quality level = MQ100
EINECS:220-570-6
UN:1224
CAS Number:2816-57-1
Formula:C₈H₁₄O
MDL Number:MFCD00001637