Order Entry
Czech Republic
ContactUsLinkComponent
1,4-Cyclohexanediol cis- and trans-mixture 99%
1,4-Cyclohexanediol cis- and trans-mixture 99%
Certificates
undefined
1,4-Cyclohexanediol cis- and trans-mixture 99%

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    97 to 102 °C
  • GC:
    ≥98.5 %
  • Iron (Fe):
    ≤20 ppm

Specification Test Results

Related Information

Frequently Bought Together