1,3-Cyclohexanediol cis- and trans-mixture ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:504-01-8
:
Formula:C₆H₁₂O₂
MDL Number:MFCD00039458
:116.16 g/mol
Boiling Pt:246…247 °C
Melting Pt:∼ 30 °C
Storage Temperature:Ambient
EINECS:207-979-5
Related Information
Frequently Bought Together