




Decahydronaphthalene (mixture of cis and trans isomers)
Katalogové číslo EHERC12093900
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pk:25 mg
- Seq:2315
Specifications
About this item
:
:138.25 g/mol
Merck Index:13,02866
Boiling Pt:189…191 °C (1013 hPa)
Melting Pt:∼–40 °C
Density:0.8696 g/cm³ (20 °C)
Flash Pt:57 °C
Storage Temperature:Ambient
EINECS:202-046-9
UN:1147
ADR:3,III
:GHS02
:GHS06
:GHS05
:GHS08
:GHS09
CAS Number:91-17-8
Formula:C₁₀H₁₈
MDL Number:MFCD00004130