ContactUsLinkComponent
Decahydronaphthalene (mixture of cis and trans isomers) cis- and trans-mixture, anhydrous 99%, AcroSeal®
Toxic
Flammable
Corrosive
Health
Environment
Decahydronaphthalene (mixture of cis and trans isomers) cis- and trans-mixture, anhydrous 99%, AcroSeal®
Certificates
undefined
Toxic
Flammable
Corrosive
Health
Environment
Decahydronaphthalene (mixture of cis and trans isomers) cis- and trans-mixture, anhydrous 99%, AcroSeal®
Specification Test Results
  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.005 %
  • GC:
    ≥98.5 % (mixture of cis and trans)