
Rhodium ≥99.9% (metals basis), sponge -20 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-16-6
:
Formula:Rh
MDL Number:MFCD00011201
ADR:4.1,III
:102.9 g/mol
EINECS:231-125-0
UN:3089
:GHS02
Storage Temperature:Ambient
Related Information
Frequently Bought Together