Platinum ≥99.99% (metals basis), slug, Premion®, Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-06-4
:
Formula:Pt
MDL Number:MFCD00011179
:195.08 g/mol
Merck Index:12,07689
Boiling Pt:3827 °C (1013 hPa)
Melting Pt:1769 °C
Density:21.447 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-116-1
Related Information
Frequently Bought Together