Order Entry
Czech Republic
ContactUsLinkComponent
Palladium ≥99.5% (trace metals basis), powder <25 nm (TEM), Sigma-Aldrich®
Irritant
Flammable
Palladium ≥99.5% (trace metals basis), powder <25 nm (TEM), Sigma-Aldrich®
Catalog # SIAL686468-500MG
Supplier:  Merck
CAS Number:  
Irritant
Flammable
Palladium ≥99.5% (trace metals basis), powder <25 nm (TEM), Sigma-Aldrich®
Catalog # SIAL686468-500MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :106.420 g/mol
Density:12.02 g/cm³ (20 °C)
ADR:4.1,II
 :M-Clarity™ quality level = MQ100
EINECS:231-115-6
 :GHS07
 :GHS02
Storage Temperature:Ambient
CAS Number:7440-05-3
Formula:Pd
MDL Number:MFCD00011167