
Palladium ≥99.9% (metals basis), powder , -60 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-05-3
:
Formula:Pd
MDL Number:MFCD00011167
:106.420 g/mol
Density:12.02 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-115-6
UN:3089
ADR:4.1,III
:GHS02
Related Information
Frequently Bought Together