Order Entry
Czech Republic
ContactUsLinkComponent
 
Palladium ≥99.9% (trace metals basis), foil, Thickness: 1.0 mm, Sigma-Aldrich®
Catalog # SIAL348678-7.6G
Supplier:  Merck
CAS Number:  
Palladium ≥99.9% (trace metals basis), foil, Thickness: 1.0 mm, Sigma-Aldrich®
Catalog # SIAL348678-7.6G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    7,6 g
  • Seq:
    3206

Specifications

About this item

 :
 :106.420 g/mol
Merck Index:12,07121
Boiling Pt:3167 °C (1013 hPa)
Melting Pt:1555 °C
Density:12.02 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:231-115-6
CAS Number:7440-05-3
Formula:Pd
MDL Number:MFCD00011167