Palladium ≥99.98% (trace metals basis), foil, Thickness: 0.25 mm, Sigma-Aldrich®
SIAL348643-1.9G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:1,9 g
- Seq:3101
:
:106.420 g/mol
:12,07121
:3167 °C (1013 hPa)
:1555 °C
:12.02 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:Ambient
:231-115-6
:7440-05-3
:Pd
:MFCD00011167