

Nickel ≥99.995% (metals basis), slug, Puratronic®, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Catalog # 42331.QK
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:25
- Seq:0025
Specifications
About this item
:
:58.69 g/mol
Merck Index:12,06582
Boiling Pt:3007 °C (1013 hPa)
Melting Pt:1453 °C
Density:8.9 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-111-4
UN:3089
ADR:4.1,III
:GHS08
:GHS07
CAS Number:7440-02-0
Formula:Ni
MDL Number:MFCD00011137