Order Entry
Czech Republic
ContactUsLinkComponent
Iridium ≥99.8% (metals basis), pellets, Ø ∼1.3 cm (0.5 in)
Iridium ≥99.8% (metals basis), pellets, Ø ∼1.3 cm (0.5 in)
Catalog # 43485.KG
CAS Number:  
Iridium ≥99.8% (metals basis), pellets, Ø ∼1.3 cm (0.5 in)
Catalog # 43485.KG
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :192.22 g/mol
Boiling Pt:5300 °C (1013 hPa)
Melting Pt:2450 °C
Storage Temperature:Ambient
EINECS:231-095-9
CAS Number:7439-88-5
Formula:Ir
MDL Number:MFCD00011062