Order Entry
Czech Republic
ContactUsLinkComponent
 
Iron ≥99.95% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 6.35 mm (0.250 in)
Catalog # 40855.KS
CAS Number:  
Iron ≥99.95% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 6.35 mm (0.250 in)
Catalog # 40855.KS
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

CAS Number:7439-89-6
 :
Formula:Fe
MDL Number:MFCD00010999
 :55.85 g/mol
Merck Index:13,05109
Boiling Pt:3000 °C (1013 hPa)
Melting Pt:1535 °C
Density:7.86 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-096-4