Manganese ≥99.95% (metals basis), irregular pieces
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7439-96-5
:
Formula:Mn
MDL Number:MFCD00011111
:54.94 g/mol
Storage Temperature:Ambient
EINECS:231-105-1
Related Information
Frequently Bought Together