
Tungsten ≥99.95% (metals basis), powder APS <1 µm
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-33-7
:
Formula:W
MDL Number:MFCD00011461
:183.84 g/mol
Storage Temperature:Ambient
EINECS:231-143-9
UN:3089
ADR:4.1,II
:GHS02
Related Information
Frequently Bought Together