Order Entry
Czech Republic
ContactUsLinkComponent
1,2-Tetradecanediol ≥90%, Technical Grade, Sigma-Aldrich®
1,2-Tetradecanediol ≥90%, Technical Grade, Sigma-Aldrich®
Catalog # SIAL260290-100G
Supplier:  Merck
CAS Number:  
1,2-Tetradecanediol ≥90%, Technical Grade, Sigma-Aldrich®
Catalog # SIAL260290-100G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :230.39 g/mol
Melting Pt:62…64 °C (lit.)
 :M-Clarity™ quality level = MQ100
CAS Number:21129-09-9
Formula:C₁₄H₃₀O₂
MDL Number:MFCD00009986