Order Entry
Czech Republic
ContactUsLinkComponent
 
Vanadium ≥99.5% (metals basis), wire, Ø 2.0 mm (0.08 in)
Catalog # 38516.G1
CAS Number:  
Vanadium ≥99.5% (metals basis), wire, Ø 2.0 mm (0.08 in)
Catalog # 38516.G1
CAS Number:  

Specifications

  • Pk:
    1 m

Specifications

About this item

 :
 :50.94 g/mol
Merck Index:12,10053
Boiling Pt:3477 °C (1013 hPa)
Melting Pt:1896 °C
Density:6.015 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-171-1
CAS Number:7440-62-2
Formula:V
MDL Number:MFCD00011453