Vanadium ≥99.8% (metals basis), wire, Ø 0.127 mm (0.005 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-62-2
:
Formula:V
MDL Number:MFCD00011453
:50.94 g/mol
Merck Index:12,10053
Boiling Pt:3477 °C (1013 hPa)
Melting Pt:1896 °C
Density:6.015 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-171-1
Frequently Bought Together