Order Entry
Czech Republic
ContactUsLinkComponent
 
Tantalum, gauze woven from wire 50 mesh, Ø 0.076 mm (0.003 in)
  40928.FL
 :  
Tantalum, gauze woven from wire 50 mesh, Ø 0.076 mm (0.003 in)
  40928.FL
 :  

 

  • Pk:
    1
  • Seq:
    1706
  • Pk Info:
    75×75 mm

 

 

 :7440-25-7
 :
 :Ta
 :MFCD00011252
 :180.95 g/mol
 :12,09223
 :5427 °C (1013 hPa)
 :2996 °C
 :16.69 g/cm³ (20 °C)
 :Ambient
 :231-135-5