Tantalum ≥99.95% (metals basis), wire, annealed, Ø 1.0 mm (0.04 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-25-7
:
Formula:Ta
MDL Number:MFCD00011252
:180.95 g/mol
Merck Index:12,09223
Boiling Pt:5427 °C (1013 hPa)
Melting Pt:2996 °C
Density:16.69 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-135-5
Frequently Bought Together