Niobium
1
SIALIRMM525A
: Merck
: 7440-03-1
- :7440-03-1
- :Nb
- :MFCD00011126
- :92.91 g/mol
- :3700 °C (1013 hPa)
- :2468 °C
- :8.57 g/cm³ (20 °C)
- :231-113-5
- Pk:1
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
#: 20864.320
: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
#: 83640.320
: VWR Chemicals
Sodium hydroxide 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM Reag. Ph. Eur. volumetric solution
#: 31770.294
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
#: 83645.320
: VWR Chemicals



