Order Entry
Czech Republic
ContactUsLinkComponent
Zirconium ≥99.95% (metals basis excluding Hf), slug (Hf ∼ 3%), Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Zirconium ≥99.95% (metals basis excluding Hf), slug (Hf ∼ 3%), Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Catalog # 42557.QK
CAS Number:  
Zirconium ≥99.95% (metals basis excluding Hf), slug (Hf ∼ 3%), Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Catalog # 42557.QK
CAS Number:  

Specifications

  • Pk:
    25
  • Seq:
    0025

Specifications

About this item

 :
 :91.22 g/mol
Storage Temperature:Ambient
EINECS:231-176-9
CAS Number:7440-67-7
Formula:Zr
MDL Number:MFCD00011303