
Titanium, powder, Sigma-Aldrich®
1.12379.0250
: Merck
:
- Pack type:Aluminium bottle
- Pk:250 g
- Seq:3545
- Solubility:(20 °C) insoluble,(reaction)
- Melting Point:1670 °C
- Molar Mass:47.88 g/mol
- Bulk Density:1260 kg/m3
- Boiling Point:3260 °C
- Density:4.5 g/cm3 (20 °C)
- pH:5 - 8 (50 g/l, H2O) suspension
- Ignition Point:300 - 450 °C
:
:47.88 g/mol
:3277 °C (1013 hPa)
:1677 °C
:4.5 g/cm³ (20 °C)
:Ambient
:231-142-3
:2546
:4.2,
:M-Clarity™ quality level = MQ200
:GHS02
:7440-32-6
:Ti
:MFCD00011264