
Titan ≥99.995% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 6.35 mm (0.250 in)
Catalog # 43045.KS
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Bal.:1
- Sekvence:0001
Specifications
About this item
:
Merck Index:12,09610
Boiling Pt:3262 °C (1013 hPa)
Melting Pt:1660 °C
Density:4,51 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-142-3
:GHS02
CAS Number:7440-32-6
Formula:Ti
MDL Number:MFCD00011264