Order Entry
Czech Republic
ContactUsLinkComponent
Titanium ≥99.7% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 6.35 mm (0.250 in)
Flammable
Titanium ≥99.7% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 6.35 mm (0.250 in)
Catalog # 40903.KS
CAS Number:  
Flammable
Titanium ≥99.7% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 6.35 mm (0.250 in)
Catalog # 40903.KS
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

CAS Number:7440-32-6
 :
Formula:Ti
MDL Number:MFCD00011264
 :47.88 g/mol
Merck Index:12,09610
Boiling Pt:3262 °C (1013 hPa)
Melting Pt:1660 °C
Density:4.51 g/cm³ (20 °C)
EINECS:231-142-3
 :GHS02
Storage Temperature:Ambient