Hafnium ≥99.7% (metals basis excluding Zr), wire (Zr ∼ 3%), Ø 2.0 mm (0.08 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-58-6
:
Formula:Hf
MDL Number:MFCD00011032
:178.49 g/mol
Storage Temperature:Ambient
EINECS:231-166-4
Frequently Bought Together