
Hafnium ≥99.6% (metals basis excluding Zr), sponge 0.8-20 mm (0.03-0.79 in) (Zr ∼ 3%)
Catalog # 00674.A1
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Typ balení:Glass bottle for solids
- Bal.:1 kg
- Sekvence:4024
Specifications
About this item
CAS Number:7440-58-6
:
Formula:Hf
MDL Number:MFCD00011032
Storage Temperature:Ambient
EINECS:231-166-4
UN:2545
ADR:4.2,II
:GHS02