Order Entry
Czech Republic
ContactUsLinkComponent
Cadmium, granular, EMSURE® ∼ 3-6 mm for analysis, Supelco®
Toxic
Health
Environment
Cadmium, granular, EMSURE® ∼ 3-6 mm for analysis, Supelco®
Catalog # 1.02004.0250
Supplier:  Merck
CAS Number:  
Toxic
Health
Environment
Cadmium, granular, EMSURE® ∼ 3-6 mm for analysis, Supelco®
Catalog # 1.02004.0250
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Metal can
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Cu (Copper):
    ≤ 0.001 %
  • Fe (Iron):
    ≤ 0.001 %
  • Hg (Mercury):
    ≤ 0.0005 %
  • Pb (Lead):
    ≤ 0.005 %
  • Zn (Zinc):
    ≤ 0.001 %

Specification Test Results

About this item

 :
 :112.41 g/mol
Merck Index:12,01649
Boiling Pt:765 °C (1013 hPa)
Melting Pt:321 °C
Density:8.6 g/cm3 (22 °C)
Storage Temperature:Ambient
EINECS:231-152-8
UN:3288
ADR:6.1,III
 :M-Clarity™ quality level = MQ300
 :GHS06
 :GHS08
 :GHS09
CAS Number:7440-43-9
Formula:Cd
MDL Number:MFCD00010914