


Cadmium ≥99.99% (metals basis), rods, Ø 12.7 mm (0.5 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-43-9
:
Formula:Cd
MDL Number:MFCD00010914
:112.41 g/mol
Merck Index:12,01649
Boiling Pt:765 °C (1013 hPa)
Melting Pt:321 °C
Density:8.642 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-152-8
UN:2570
ADR:6.1,II
:GHS06
:GHS08
:GHS09
Frequently Bought Together