Order Entry
Czech Republic
ContactUsLinkComponent
Cadmium ≥99.95% (metals basis), foil 15x15 cm (5.9x5.9 in), Thickness: 1.0 mm (0.04 in)
Toxic
Health
Environment
Cadmium ≥99.95% (metals basis), foil 15x15 cm (5.9x5.9 in), Thickness: 1.0 mm (0.04 in)
Catalog # 00940.KF
CAS Number:  
Toxic
Health
Environment
Cadmium ≥99.95% (metals basis), foil 15x15 cm (5.9x5.9 in), Thickness: 1.0 mm (0.04 in)
Catalog # 00940.KF
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001
  • Pk Info:
    1 piece

Specifications

About this item

 :
 :112.41 g/mol
Merck Index:12,01649
Boiling Pt:765 °C (1013 hPa)
Melting Pt:321 °C
Density:8.642 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-152-8
UN:2570
ADR:6.1,II
 :GHS06
 :GHS08
 :GHS09
CAS Number:7440-43-9
Formula:Cd
MDL Number:MFCD00010914