Order Entry
Czech Republic
ContactUsLinkComponent
Silver, Sigma-Aldrich®
Silver, Sigma-Aldrich®
Catalog # SIAL265551-330MG
Supplier:  Merck
CAS Number:  
Silver, Sigma-Aldrich®
Catalog # SIAL265551-330MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    330 mg
  • Seq:
    2635

Specifications

About this item

 :
 :107.87 g/mol
Merck Index:12,08647
Boiling Pt:2212 °C (1013 hPa)
Melting Pt:962 °C
Density:10.5 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-131-3
CAS Number:7440-22-4
Formula:Ag
MDL Number:MFCD00003397