Silver ≥99.999% (metals basis), shot, Premion® 2-3 mm (0.08-0.12 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-22-4
:
Formula:Ag
MDL Number:MFCD00003397
:107.87 g/mol
Merck Index:12,08647
Boiling Pt:2212 °C (1013 hPa)
Melting Pt:962 °C
Density:10.5 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-131-3
Related Information
Frequently Bought Together