Order Entry
Czech Republic
ContactUsLinkComponent
Silver hard ≥99.99% (metals basis), rods, Premion®, hard, Length: 150 mm (5.9 in), Ø 6.35 mm (0.25 in)
Silver hard ≥99.99% (metals basis), rods, Premion®, hard, Length: 150 mm (5.9 in), Ø 6.35 mm (0.25 in)
Catalog # 13304.KG
CAS Number:  
Silver hard ≥99.99% (metals basis), rods, Premion®, hard, Length: 150 mm (5.9 in), Ø 6.35 mm (0.25 in)
Catalog # 13304.KG
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

CAS Number:7440-22-4
 :
Formula:Ag
MDL Number:MFCD00003397
 :107.87 g/mol
Merck Index:12,08647
Boiling Pt:2212 °C (1013 hPa)
Melting Pt:962 °C
Density:10.5 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-131-3